4-Phenoxypiperidine ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

H50984.03 H50984.MD
H50984.03EA 106 GBP
H50984.03 H50984.MD
4-Phenoxypiperidine ≥99%
4-Phenoxypiperidine

Purity: 99% CAS No.: 3202-33-3 Molecular Formula: C11H15NO Formula Weight: 177.25 Boiling Point: 107-108°/0.1mm MDL No.: MFCD04114971

Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
Boiling Pt: 107…108 °C (0.1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD04114971
CAS Number: 3202-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR