1-Boc-4-(4-aminophenoxy)piperidine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2643-5G
HC-2643-1G
HC-2643-250M
COBBHC-2643-5GEA
0
GBP
COBBHC-2643-5G
COBBHC-2643-1G
COBBHC-2643-250M
1-Boc-4-(4-aminophenoxy)piperidine
1-Boc-4-(4-aminophenoxy)piperidine
Formula:
C₁₆H₂₄N₂O₃ |
MDL Number:
MFCD04115023 CAS Number: 138227-63-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...