1-(2-Pyridin-2-ylethyl)piperazine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

019054-5G 019054-1G
FLUO019054-5GEA 0 GBP
FLUO019054-5G FLUO019054-1G
1-(2-Pyridin-2-ylethyl)piperazine
1-(2-Pyridin-2-ylethyl)piperazine
Formula: C₁₁H₁₇N₃
MDL Number: MFCD01748645
CAS Number: 53345-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR