2-Methoxy-5-(tributylstannyl)thiazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51223G1 AS51223G5
ACTIAS51223G1EA 197 GBP
ACTIAS51223G1 ACTIAS51223G5
2-Methoxy-5-(tributylstannyl)thiazole
2-Methoxy-5-(tributylstannyl)thiazole
Formula: C₁₆H₃₁NOSSn
MDL Number: MFCD09025806
CAS Number: 1025744-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR