2-Ethoxy-5-(tributylstannyl)thiazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51222G1 AS51222G5
ACTIAS51222G1EA 0 GBP
ACTIAS51222G1 ACTIAS51222G5
2-Ethoxy-5-(tributylstannyl)thiazole
2-Ethoxy-5-(tributylstannyl)thiazole
Formula: C₁₇H₃₃NOSSn
MDL Number: MFCD09025788
CAS Number: 446285-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR