2-Chloro-5-(tributylstannyl)thiazole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS50263G1 AS50263G5
ACTIAS50263G1EA 0 GBP
ACTIAS50263G1 ACTIAS50263G5
2-Chloro-5-(tributylstannyl)thiazole
2-Chloro-5-(tributylstannyl)thiazole
Formula: C₁₅H₂₈ClNSSn
MDL Number: MFCD09265501
CAS Number: 889672-73-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR