1-Methyl-2-(tributylstannyl)pyrrole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS50215G1 AS50215G5
ACTIAS50215G1EA 0 GBP
ACTIAS50215G1 ACTIAS50215G5
1-Methyl-2-(tributylstannyl)pyrrole
1-Methyl-2-(tributylstannyl)pyrrole
Formula: C₁₇H₃₃NSn
MDL Number: MFCD01319022
CAS Number: 118486-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR