5-(Tributylstannyl)pyrimidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51163G1 AS51163G5
ACTIAS51163G1EA 0 GBP
ACTIAS51163G1 ACTIAS51163G5
5-(Tributylstannyl)pyrimidine
5-(Tributylstannyl)pyrimidine
Formula: C₁₆H₃₀N₂Sn
MW: 369.13 g/mol
MDL Number: MFCD01319056
CAS Number: 144173-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR