2-(Methylthio)-5-(tributylstannyl)pyrimidine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51169G1 AS51169G5
ACTIAS51169G1EA 0 GBP
ACTIAS51169G1 ACTIAS51169G5
2-(Methylthio)-5-(tributylstannyl)pyrimidine
2-(Methylthio)-5-(tributylstannyl)pyrimidine
Formula: C₁₇H₃₂N₂SSn
MDL Number: MFCD07787371
CAS Number: 120717-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR