4-(Tributylstannyl)pyridazine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

AS51166G1 AS51166G5
ACTIAS51166G1EA 0 GBP
ACTIAS51166G1 ACTIAS51166G5
4-(Tributylstannyl)pyridazine
4-(Tributylstannyl)pyridazine
Formula: C₁₆H₃₀N₂Sn
MDL Number: MFCD07787369
CAS Number: 194865-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR