Cyclopentadienylmolybdenum tricarbonyl dimer ≥98%

Supplier: Thermo Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: Bis(tricarbonylcyclopentadienylmolybdenum)

39245.03 39245.06 39245.14
39245.03EA 43.8 GBP
39245.03 39245.06 39245.14
Cyclopentadienylmolybdenum tricarbonyl dimer ≥98%
Cyclopentadienylmolybdenum tricarbonyl dimer

Purity: 98% CAS No.: 12091-64-4 Molecular Formula: C16H10Mo2O6 Formula Weight: 490.13 Physical Form: Crystalline Sensitivity: Air Sensitive Appearance: Purple to red Melting Point: 220° dec. UN No.: 3466 MDL No.: MFCD00009947

Formula: C₁₆H₁₀Mo₂O₆
MW: 490.13 g/mol
Melting Pt: 220 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00009947
CAS Number: 12091-64-4
EINECS: 235-156-0
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR