3-(Cyclohexylcarbamoyl)-5-fluorophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2857-5G BB-2857-1G
COBBBB-2857-5GEA 0 GBP
COBBBB-2857-5G COBBBB-2857-1G
3-(Cyclohexylcarbamoyl)-5-fluorophenylboronic acid
3-(Cyclohexylcarbamoyl)-5-fluorophenylboronic acid
Formula: C₁₃H₁₇BFNO₃
MDL Number: MFCD08235059
CAS Number: 874219-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR