3-(2-Aminoethoxy)benzonitrile

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22874G1 AS22874G5
ACTIAS22874G1EA 167 GBP
ACTIAS22874G1 ACTIAS22874G5
3-(2-Aminoethoxy)benzonitrile
3-(2-Aminoethoxy)benzonitrile
Formula: C₉H₁₀N₂O
MW: 162.1900 g/mol
MDL Number: MFCD16495888
CAS Number: 120351-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR