3-(Cyclohexyl(methyl)carbamoyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-3854-5G BB-3854-1G
COBBBB-3854-5GEA 0 GBP
COBBBB-3854-5G COBBBB-3854-1G
3-(Cyclohexyl(methyl)carbamoyl)phenylboronic acid
3-(Cyclohexyl(methyl)carbamoyl)phenylboronic acid
Formula: C₁₄H₂₀BNO₃
MDL Number: MFCD09972095
CAS Number: 1072945-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR