3,5-Dichloropyridine-2-carbonitrile
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3,5-Dichloropicolinonitrile, 3,5-Dichloro-2-cyanopyridine
OS-7674-25G
OS-7674-100G
OS-7674-5G
COBBOS-7674-25GEA
0
GBP
COBBOS-7674-25G
COBBOS-7674-100G
COBBOS-7674-5G
3,5-Dichloropyridine-2-carbonitrile
3,5-Dichloropyridine-2-carbonitrile
Formula:
C₆H₂Cl₂N₂ MW: 173.00 g/mol |
MDL Number:
MFCD03788758 CAS Number: 85331-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...