4-Chloro-3-(cyclohexylcarbamoyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2844-5G BB-2844-1G
COBBBB-2844-5GEA 0 GBP
COBBBB-2844-5G COBBBB-2844-1G
4-Chloro-3-(cyclohexylcarbamoyl)phenylboronic acid
4-Chloro-3-(cyclohexylcarbamoyl)phenylboronic acid
Formula: C₁₃H₁₇BClNO₃
MDL Number: MFCD07783884
CAS Number: 871332-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR