4-(Cyclohexylcarbamoyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Cyclohexylaminocarbonyl)phenylboronic acid

BB-3015-1G BB-3015-5G BB-3015-25G
COBBBB-3015-1GEA 0 GBP
COBBBB-3015-1G COBBBB-3015-5G COBBBB-3015-25G
4-(Cyclohexylcarbamoyl)phenylboronic acid
4-(Cyclohexylcarbamoyl)phenylboronic acid
Formula: C₁₃H₁₈BNO₃
MDL Number: MFCD03411956
CAS Number: 762262-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR