4-(Methoxycarbonyl)phenylboronic acid pinacol ester

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-(Methoxycarbonyl)benzeneboronic acid pinacol ester, Methyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

010813-5G 010813-1G 010813-25G
FLUO010813-5GEA 0 GBP
FLUO010813-5G FLUO010813-1G FLUO010813-25G
4-(Methoxycarbonyl)phenylboronic acid pinacol ester
4-(Methoxycarbonyl)phenylboronic acid pinacol ester
Formula: C₁₄H₁₉BO₄
MW: 262.11 g/mol
MDL Number: MFCD02179438
CAS Number: 171364-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR