2-(Trifluoromethyl)-5-pyridineboronic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 6-(Trifluoromethyl)-3-pyridylboronic acid, 2-(Trifluoromethyl)pyridin-5-ylboronic acid, 2-(Trifluoromethyl)pyridine-5-boronic acid, (6-(Trifluoromethyl)pyridin-3-yl)boronic acid

AS22989G1 AS22989G5
ACTIAS22989G1EA 0 GBP
ACTIAS22989G1 ACTIAS22989G5
2-(Trifluoromethyl)-5-pyridineboronic acid
2-(Trifluoromethyl)-5-pyridineboronic acid
Formula: C₆H₅BF₃NO₂
MW: 190.92 g/mol
MDL Number: MFCD07375381
CAS Number: 868662-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR