1-Cyclohexenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Cyclohex-1-en-1-ylboronic acid
BB-2511-1G
BB-2511-5G
COBBBB-2511-1GEA
0
GBP
COBBBB-2511-1G
COBBBB-2511-5G
1-Cyclohexenylboronic acid
1-Cyclohexenylboronic acid
Formula:
C₆H₁₁BO₂ MW: 125.96 g/mol |
MDL Number:
MFCD02179497 CAS Number: 89490-05-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...