3-Bromo-N-hexyl-5-(trifluoromethyl)benzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Bromo-5-(N-hexylsulphamoyl)benzotrifluoride

HC-6302-5G HC-6302-1G
COBBHC-6302-5GEA 0 GBP
COBBHC-6302-5G COBBHC-6302-1G
3-Bromo-N-hexyl-5-(trifluoromethyl)benzenesulphonamide
3-Bromo-N-hexyl-5-(trifluoromethyl)benzenesulphonamide
Formula: C₁₃H₁₇BrF₃NO₂S
MDL Number: MFCD09801008
CAS Number: 951884-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR