3-Bromo-N,N-dimethyl-5-(trifluoromethyl)benzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6308-5G HC-6308-1G
COBBHC-6308-5GEA 0 GBP
COBBHC-6308-5G COBBHC-6308-1G
3-Bromo-N,N-dimethyl-5-(trifluoromethyl)benzenesulphonamide
3-Bromo-N,N-dimethyl-5-(trifluoromethyl)benzenesulphonamide
Formula: C₉H₉BrF₃NO₂S
MDL Number: MFCD09801011
CAS Number: 951884-65-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR