3-Isoquinolinol

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Hydroxyisoquinoline

AS21956G1 AS21956G25 AS21956G5
ACTIAS21956G1EA 14 GBP
ACTIAS21956G1 ACTIAS21956G25 ACTIAS21956G5
3-Isoquinolinol
3-Isoquinolinol
Formula: C₉H₇NO
MW: 145.16 g/mol
MDL Number: MFCD00075524
CAS Number: 7651-81-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR