1-Bromo-3-fluoro-4-nitrobenzene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-2-fluoronitrobenzene

AS21633G100 AS21633G25 AS21633G5
ACTIAS21633G100EA 0 GBP
ACTIAS21633G100 ACTIAS21633G25 ACTIAS21633G5
1-Bromo-3-fluoro-4-nitrobenzene
1-Bromo-3-fluoro-4-nitrobenzene
Formula: C₆H₃BrFNO₂
MDL Number: MFCD01930221
CAS Number: 321-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR