(2,3-Difluorophenyl)acetic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-(2,3-Difluorophenyl)acetic acid

AS21910G100 AS21910G25 AS21910G5
ACTIAS21910G100EA 0 GBP
ACTIAS21910G100 ACTIAS21910G25 ACTIAS21910G5
(2,3-Difluorophenyl)acetic acid
(2,3-Difluorophenyl)acetic acid
Formula: (C₆H₃F₂)CH₂COOH
MW: 172.13 g/mol
MDL Number: MFCD00040968
CAS Number: 145689-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR