(3-Hydroxy-4-methoxyphenyl)acetic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-(3-Hydroxy-4-methoxyphenyl)acetic acid
AS20540G1
AS20540G25
AS20540G5
ACTIAS20540G1EA
0
GBP
ACTIAS20540G1
ACTIAS20540G25
ACTIAS20540G5
(3-Hydroxy-4-methoxyphenyl)acetic acid
(3-Hydroxy-4-methoxyphenyl)acetic acid
Formula:
(C₆H₃(OH)(OCH₃))CH₂COOH MW: 182.18 g/mol |
MDL Number:
MFCD00016829 CAS Number: 1131-94-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...