2-Isobutoxy-5-(trifluoromethyl)-3-pyridineboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-6238-5G BB-6238-1G
COBBBB-6238-5GEA 0 GBP
COBBBB-6238-5G COBBBB-6238-1G
2-Isobutoxy-5-(trifluoromethyl)-3-pyridineboronic acid
2-Isobutoxy-5-(trifluoromethyl)-3-pyridineboronic acid
Formula: C₁₀H₁₃BF₃NO₃
MDL Number: MFCD15143472
CAS Number: 1218790-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR