2-Methoxy-6-(trifluoromethyl)-4-pyridineboronic acid pinacol ester
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PN-5769-1G
PN-5769-250M
COBBPN-5769-1GEA
0
GBP
COBBPN-5769-1G
COBBPN-5769-250M
2-Methoxy-6-(trifluoromethyl)-4-pyridineboronic acid pinacol ester
2-Methoxy-6-(trifluoromethyl)-4-pyridineboronic acid pinacol ester
Formula:
C₁₃H₁₇BF₃NO₃ |
MDL Number:
MFCD12026100 CAS Number: 1150561-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...