5-Methyl-3-pyridineboronic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Methylpyridine-3-boronic acid

AS50994G1 AS50994G5
ACTIAS50994G1EA 27 GBP
ACTIAS50994G1 ACTIAS50994G5
5-Methyl-3-pyridineboronic acid
5-Methyl-3-pyridineboronic acid
Formula: C₆H₈BNO₂
MW: 136.95 g/mol
MDL Number: MFCD03428528
CAS Number: 173999-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR