5-Fluoro-2-methoxy-4-pyridineboronic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Fluoro-2-methoxypyridine-4-boronic acid

AS31128G1 AS31128G5
ACTIAS31128G1EA 0 GBP
ACTIAS31128G1 ACTIAS31128G5
5-Fluoro-2-methoxy-4-pyridineboronic acid
5-Fluoro-2-methoxy-4-pyridineboronic acid
Formula: C₆H₇BFNO₃
MDL Number: MFCD06657887
CAS Number: 1043869-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR