5-Methoxypyridine-3-boronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-methoxypyridin-3-yl boronic acid, (5-Methoxypyridin-3-yl)boronic acid, 5-Methoxy-3-pyridineboronic acid

BB-5109-1G BB-5109-5G
COBBBB-5109-1GEA 0 GBP
COBBBB-5109-1G COBBBB-5109-5G
5-Methoxypyridine-3-boronic acid
5-Methoxypyridine-3-boronic acid
Formula: C₆H₈BNO₃
MW: 152.95 g/mol
MDL Number: MFCD03425977
CAS Number: 850991-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR