Methyl-3-bromo-5-(trifluoromethyl)benzoate
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4592-5G
CA-4592-25G
CA-4592-1G
COBBCA-4592-5GEA
0
GBP
COBBCA-4592-5G
COBBCA-4592-25G
COBBCA-4592-1G
Methyl-3-bromo-5-(trifluoromethyl)benzoate
Methyl 3-bromo-5-(trifluoromethyl)benzoate
Formula:
C₉H₆BrF₃O₂ |
MDL Number:
MFCD11847338 CAS Number: 187331-46-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...