5-Fluoro-2-(methylamino)benzotrifluoride hydrochloride

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-2880-1G HC-2880-5G
COBBHC-2880-1GEA 0 GBP
COBBHC-2880-1G COBBHC-2880-5G
5-Fluoro-2-(methylamino)benzotrifluoride hydrochloride
5-Fluoro-2-(methylamino)benzotrifluoride hydrochloride
Formula: C₈H₈ClF₄N
MDL Number: MFCD13195690
CAS Number: 1215205-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR