2-Fluoro-5-(methylamino)benzotrifluoride hydrochloride
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2875-5G
HC-2875-1G
COBBHC-2875-5GEA
0
GBP
COBBHC-2875-5G
COBBHC-2875-1G
2-Fluoro-5-(methylamino)benzotrifluoride hydrochloride
2-Fluoro-5-(methylamino)benzotrifluoride hydrochloride
Formula:
C₈H₈ClF₄N |
MDL Number:
MFCD12913960 CAS Number: 1187386-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...