Methyl-4-amino-3-fluorobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS35126G1 AS35126G5
ACTIAS35126G1EA 0 GBP
ACTIAS35126G1 ACTIAS35126G5
Methyl-4-amino-3-fluorobenzoate
Methyl 4-amino-3-fluorobenzoate
Formula: C₈H₈FNO₂
MDL Number: MFCD08689703
CAS Number: 185629-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR