Methyl-4-amino-3-chlorobenzoate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 4-amino-3-chlorobenzoate

012690-25G 012690-5G
FLUO012690-25GEA 0 GBP
FLUO012690-25G FLUO012690-5G
Methyl-4-amino-3-chlorobenzoate
Methyl 4-amino-3-chlorobenzoate
Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
MDL Number: MFCD01320686
CAS Number: 84228-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR