Methyl-4-amino-2-chlorobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS22319G25 AS22319G5
ACTIAS22319G25EA 0 GBP
ACTIAS22319G25 ACTIAS22319G5
Methyl-4-amino-2-chlorobenzoate
Methyl 4-amino-2-chlorobenzoate
Formula: C₈H₈ClNO₂
MDL Number: MFCD03840484
CAS Number: 46004-37-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR