Methyl-3-amino-5-chlorobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 3-amino-5-chlorobenzoate

AS23458G1 AS23458G5
ACTIAS23458G1EA 0 GBP
ACTIAS23458G1 ACTIAS23458G5
Methyl-3-amino-5-chlorobenzoate
Methyl 3-amino-5-chlorobenzoate
Formula: C₈H₈ClNO₂
MW: 185.6076 g/mol
MDL Number: MFCD11053764
CAS Number: 21961-31-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR