Ethyl-3-amino-5-chlorobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23632G1 AS23632G5
ACTIAS23632G1EA 184 GBP
ACTIAS23632G1 ACTIAS23632G5
Ethyl-3-amino-5-chlorobenzoate
Ethyl 3-amino-5-chlorobenzoate
Formula: C₉H₁₀ClNO₂
MDL Number: MFCD17276491
CAS Number: 1261269-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR