2-Bromo-N-(2-methylphenyl)benzamide
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CS-9434-5G
CS-9434-1G
COBBCS-9434-5GEA
0
GBP
COBBCS-9434-5G
COBBCS-9434-1G
2-Bromo-N-(2-methylphenyl)benzamide
2-Bromo-N-(2-methylphenyl)benzamide
Formula:
C₁₄H₁₂BrNO |
MDL Number:
MFCD00783656 CAS Number: 303991-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...