6-Bromo-1-chloroisoquinoline
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS2693G5
AS2693G1
ACTIAS2693G5EA
110
GBP
ACTIAS2693G5
ACTIAS2693G1
6-Bromo-1-chloroisoquinoline
6-Bromo-1-chloroisoquinoline
Formula:
C₉H₅BrClN MW: 242.5000 g/mol |
MDL Number:
MFCD06738661 CAS Number: 205055-63-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...