4-Bromo-1-chloroisoquinoline

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS2039G1 AS2039G5
ACTIAS2039G1EA 0 GBP
ACTIAS2039G1 ACTIAS2039G5
4-Bromo-1-chloroisoquinoline
4-Bromo-1-chloroisoquinoline
Formula: C₉H₅BrClN
MDL Number: MFCD00234469
CAS Number: 66728-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR