1-Chloro-6-methylisoquinoline
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS21106G1
AS21106G5
ACTIAS21106G1EA
0
GBP
ACTIAS21106G1
ACTIAS21106G5
1-Chloro-6-methylisoquinoline
1-Chloro-6-methylisoquinoline
Formula:
C₁₀H₈ClN |
MDL Number:
MFCD11110400 CAS Number: 209286-73-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...