Methyl-3-aminoisonicotinate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 3-aminoisonicotinate, Methyl 3-aminopyridine-4-carboxylate
AS22708G1
AS22708G5
ACTIAS22708G1EA
13
GBP
ACTIAS22708G1
ACTIAS22708G5
Methyl-3-aminoisonicotinate
Methyl 3-aminoisonicotinate
Formula:
C₇H₈N₂O₂ MW: 152.20 g/mol |
MDL Number:
MFCD02082535 CAS Number: 55279-30-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...