Methyl-2-bromoisonicotinate, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2-bromoisonicotinate

M-Clarity™ quality level = MQ100

689505-1G 689505-5G
SIAL689505-1GEA 129 GBP
SIAL689505-1G SIAL689505-5G
Methyl-2-bromoisonicotinate, Sigma-Aldrich®
Methyl 2-bromoisonicotinate
Formula: C₇H₆BrNO₂
MW: 216.0320 g/mol
MDL Number: MFCD03791265
CAS Number: 26156-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR