Methyl-2-bromoisonicotinate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2-bromoisonicotinate

CA-4198-5G CA-4198-25G CA-4198-1G
COBBCA-4198-5GEA 0 GBP
COBBCA-4198-5G COBBCA-4198-25G COBBCA-4198-1G
Methyl-2-bromoisonicotinate
Methyl 2-bromoisonicotinate
Formula: C₇H₆BrNO₂
MW: 216.0320 g/mol
MDL Number: MFCD03791265
CAS Number: 26156-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR