Methyl-4-fluoro-3-methylbenzoate
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
Methyl 4-fluoro-3-methylbenzoate
AS22816G25
AS22816G5
ACTIAS22816G25EA
138
GBP
ACTIAS22816G25
ACTIAS22816G5
Methyl-4-fluoro-3-methylbenzoate
Methyl 4-fluoro-3-methylbenzoate
Formula:
C₉H₉FO₂ MW: 168.1650 g/mol |
MDL Number:
MFCD06203783 CAS Number: 180636-50-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...