Methyl-2-fluoro-6-methylbenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 2-fluoro-6-methylbenzoate

AS24925G1 AS24925G5
ACTIAS24925G1EA 32 GBP
ACTIAS24925G1 ACTIAS24925G5
Methyl-2-fluoro-6-methylbenzoate
Methyl 2-fluoro-6-methylbenzoate
Formula: C₉H₉FO₂
MW: 168.1700 g/mol
MDL Number: MFCD11226310
CAS Number: 197516-57-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR