Methyl-4-bromo-2-methylbenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 4-bromo-2-methylbenzoate

AS22796G25 AS22796G5
ACTIAS22796G25EA 0 GBP
ACTIAS22796G25 ACTIAS22796G5
Methyl-4-bromo-2-methylbenzoate
Methyl 4-bromo-2-methylbenzoate
Formula: C₉H₉BrO₂
MW: 229.0706 g/mol
MDL Number: MFCD09954957
CAS Number: 99548-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR