Methyl-3-bromo-2-fluorobenzoate

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 3-bromo-2-fluorobenzoate

AS34530G1 AS34530G5
ACTIAS34530G1EA 0 GBP
ACTIAS34530G1 ACTIAS34530G5
Methyl-3-bromo-2-fluorobenzoate
Methyl 3-bromo-2-fluorobenzoate
Formula: C₈H₆BrFO₂
MDL Number: MFCD09261255
CAS Number: 206551-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR